Learn More
Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, human, non-targeting, with GFP
Negative control gRNAs are essential in screening applications for setting thresholds for hit determination.
£382.00 - £1526.00
Specifications
Delivery Type | Lentiviral |
---|---|
Format | Tube |
Control Type | Negative Control |
RNAi Type | gRNA |
Species | Human |
Alert:
For Research Use Only. Not for use in diagnostic procedures.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15607898
|
Invitrogen™
A32063 |
100 mL |
£382.00
100µL |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920383
|
Invitrogen™
A32831 |
1 mL |
£1526.00
1mL |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Negative control gRNAs are essential in screening applications for setting thresholds for hit determination.
The Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, Human Scrambled, contains a gRNA sequence with no sequence homology to any region of the human genome. This construct also expresses emGFP to provide a visual readout of successful transduction that can be used in determining Multiplicity of Infection (MOI).
The Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, Human Scrambled, contains a gRNA sequence with no sequence homology to any region of the human genome. This construct also expresses emGFP to provide a visual readout of successful transduction that can be used in determining Multiplicity of Infection (MOI).
Specifications
Lentiviral | |
Negative Control | |
Human | |
LentiArray™ | |
U6 |
Tube | |
gRNA | |
Genome Editing | |
gRNA | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.