missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, human, non-targeting
Brand: Invitrogen™ A32327
Alert:
For Research Use Only. Not for use in diagnostic procedures.
Description
Negative control gRNAs are essential in screening applications for setting thresholds for hit determination.
The Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, Human Scrambled, contains a gRNA sequence with no sequence homology to any region of the human genome.
Recommended Storage
Store at -68 to -85°C
Specifications
1 x 1 mL. Store at -68 to -85°C. | |
Negative Control | |
Human | |
LentiArray™ | |
1 mL |
Tube | |
gRNA | |
Genome Editing | |
gRNA | |
Dry Ice |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction