Learn More
Invitrogen™ LentiArray™ CRISPR Positive Control Lentivirus, human HPRT, with GFP
Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system.
£382.00 - £1526.00
Specifications
Delivery Type | Lentiviral |
---|---|
Format | Tube |
Control Type | Positive Control |
RNAi Type | gRNA |
Species | Human |
Alert:
For Research Use Only. Not for use in diagnostic procedures.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15697888
|
Invitrogen™
A32060 |
100 mL |
£382.00
100µL |
Estimated Shipment: 25-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15910383
|
Invitrogen™
A32830 |
1 mL |
£1526.00
1mL |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Optimizing delivery conditions is a critical step in any experiment utilizing the CRISPR/Cas9 system. The Invitrogen™ LentiArray CRISPR Positive Control Lentivirus gRNA against HPRT is a highly efficient gRNA validated in multiple human cell lines that has achieved editing efficiencies of greater than 80%.
The Invitrogen™ LentiArray CRISPR Positive Control Lentivirus gRNA against HPRT is a highly efficient gRNA validated in multiple human cell lines that has achieved editing efficiencies of greater than 80%. It can be used to rapidly determine ideal delivery conditions in order to achieve maximum editing efficiency in your cell model of choice. This construct also expresses emGFP to provide a visual readout of successful transduction that can be used in determining Multiplicity of Infection (MOI).
Specifications
Lentiviral | |
Positive Control | |
Human | |
LentiArray™ | |
U6 |
Tube | |
gRNA | |
Genome Editing | |
gRNA | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.