missing translation for 'onlineSavingsMsg'
Learn More

LentiArray™ LentiArray™ Human Nuclear Hormone Receptor CRISPR Library, Glycerol

Product Code. 15920363
Click to view available options
:
1 library
This item is not returnable. View return policy

Product Code. 15920363

Brand: LentiArray™ A32175

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Targets 47 genes with up to 4 gRNA per gene target for a total of 188 gRNAs. Libraries are delivered as 50μL of glycerol stock per gRNA and are also available as ready to use packaged lentivirus.

Nuclear hormone receptors are a family of ligand-activated transcription factors that are activated by lipid-soluble ligands such as steroid hormones, thyroid hormone, vitamin D, and retinoic acid. Nuclear hormone receptors regulate a range of biological processes, including metabolism, development, proliferation, and reproduction. The gene targets within this library were selected using the most up-to-date genome databases, including the NCBI RefSeq database, and cross-referenced to the Gene Ontology Consortium (GO) database and/or the HUGO Gene Nomenclature Committee (HGNC).

Built to bring you success
 LentiArray CRISPR libraries are constructed using gRNA designs produced using Thermo Fisher Scientific’s proprietary CRISPR gRNA design algorithm. This algorithm incorporates the latest gRNA design research, as well as our extensive in-house experience, to produce the highest quality designs. The gRNA designs included in the LentiArray CRISPR libraries are selected for maximal editing efficiency and specificity and are designed to knock out all known isoforms of the target gene. For each gene target, up to four high-quality gRNAs are included to help ensure highly efficient knockout of the target gene in a wide array of cell types.

Design and execute your experiments without limitations
  • LentiArray CRISPR libraries are designed and constructed to give you complete control over your experimental design. They utilize a two-vector design, expressing the Cas9 nuclease and the gRNA off of separate lentiviral constructs, enabling you to dictate when and how the genome editing tools are delivered to your cells.
  • The LentiArray Cas9 lentivirus construct expresses a human, codon-optimized Cas9 nuclease with a blasticidin-resistance gene under the control of the EF-1α (EFS) promoter. You can choose to co-infect your cells with both the LentiArray gRNA lentiviruses and the Cas9 lentivirus or just the LentiArray Cas9 lentivirus to establish a stable Cas9 expressing cell line. With LentiArray CRISPR libraries you have the freedom to design the experimental approach that best suits your model system and your screening goals.
  • Furthermore, to minimize restrictions to your experimental design, the gRNA lentiviruses that make up the LentiArray CRISPR libraries do not contain fluorescent markers. This allows reporters of any wavelength to be used with the libraries and expands your ability to perform multiplexed experiments. For challenging cell lines, the LentiArray gRNA lentiviruses contain a puromycin resistance gene that allows enrichment of the population of edited cells and drives stronger phenotypes.

Recommended Storage

Store at −68° to −85°C.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 library set, 50 μL/target.
Store at -68 to -85°C.
For Use With (Application) Genome Editing
Product Line LentiArray
Product Type Crispr Library
Quantity 1 Library
Shipping Condition Dry Ice
Format 96-well Plate
RNAi Type gRNA
Sample Type DNA
Species Human

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.