Learn More
LentiArray™ LentiArray™ Human Tumor Suppressor Library, Glycerol Stock
Targets 716 genes with up to 4 gRNA per gene target for a total of 2,864 gRNAs. Libraries are delivered as 50μL of glycerol stock per gRNA and are also available as ready to use packaged lentivirus
Brand:  LentiArray™ A32181
Product Code. 15980363
  • £17654.20 / Each
Explore more special offers
Description
Show More
Specifications
Specifications
Glycerol Stock
1 Library
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
LentiArray™ LentiArray™ Human Tumor Suppressor Library, Glycerol Stock > 1 library