missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emma Safety Footwear Leo All-Round Safety D-Fit Shoes
Designed for people with slightly broader-than-normal feet.
£104.00 - £127.00
Specifications
Liner Material | Hydro-Tec Sanitized Silver lining |
---|---|
Sole Features | Non-woven antistatic insole |
Sole Material | GripForce™ Easy Twist outsole |
Product Type | All-Round Safety Shoes |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
10012104
|
Emma Safety Footwear
MM70054844 |
44 |
£127.00
1 pair |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||||
10464907
|
Emma Safety Footwear
MM70054847 |
47 |
£104.00
1 pair |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||||
10431714
|
Emma Safety Footwear
MM70054842 |
42 |
£104.00
1 pair |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||||
10215404
|
Emma Safety Footwear
MM70054841 |
41 |
£104.00
1 pair |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||||
10738675
|
Emma Safety Footwear
MM70054843 |
43 |
£104.00
1 pair |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||||
10379597
|
Emma Safety Footwear
MM70054840 |
40 |
£104.00
1 pair |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||||
10286264
|
Emma Safety Footwear
MM70054845 |
45 |
£104.00
1 pair |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||||
10022104
|
Emma Safety Footwear
MM70054846 |
46 |
£104.00
1 pair |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||||
Description
- Full grain leather uppers
- Hydro-Tec™ sanitized silver lining
- Non-woven antistatic insole
- GripForce™ Easy Twist outsole
- Strobel-stitched upper construction
Specifications
Hydro-Tec Sanitized Silver lining | |
Non-woven antistatic insole | |
GripForce™ Easy Twist outsole | |
All-Round Safety Shoes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Emma Safety Footwear Leo All-Round Safety D-Fit Shoes