Learn More
Greiner Bio-One Leucosep Polypropylene Round Bottom Centrifugation Tubes
For efficient separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow.
£466.00
Specifications
Material | Polypropylene |
---|---|
For Use With | Separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow |
Includes | Porous barrier |
Bottom Shape | Round |
Product Code | Brand | Capacity (Metric) | Product Type | Sterility | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Product Type | Sterility | Price | Quantity & Availability | |||
17245314
|
Greiner Bio-One
227288 |
50 mL | Centrifuge Tube | Aseptically Manufactured |
£466.00
Pack of 25 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
Description
- Enrichment directly from whole blood
- Simplified filling through porous barrier
- Rapid separation in 15 minutes at room temperature
- Removal of erythrocytes and granulocytes
- No recontamination with erythrocytes
Service Plan
No
Specifications
Polypropylene | |
Separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow | |
Porous barrier | |
Round |