missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leukotriene B4, Tocris Bioscience™
Click to view available options
Quantity:
50 μg
Description
Potent lipid inflammatory mediator derived from the 5-lipoxygenase pathway of arachidonic acid metabolism. Binds to BLT1 and BLT2 receptors and acts as a potent chemotactic agent and activator of leukocytes. Also displays antiviral activity towards DNA viruses and retroviruses.
Specifications
Specifications
CAS | 71160-24-2 |
Molecular Formula | C20H32O4 |
Quantity | 50 μg |
Synonym | leukotriene b4, ltb4, 5,12-dihete, 5,12-hete, unii-1hgw4dr56d, 5s,12r-dihete, 1hgw4dr56d, 5s,6z,8e,10e,12r,14z-5,12-dihydroxyicosa-6,8,10,14-tetraenoic acid, chembl65061, 5s,12r-dihydroxy-6z,8e,10e,14z-eicosatetraenoic acid |
InChI Key | VNYSSYRCGWBHLG-AMOLWHMGSA-N |
SMILES | CCCCCC=CCC(C=CC=CC=CC(CCCC(=O)O)O)O |
IUPAC Name | (5S,6Z,8E,10E,12R,14Z)-5,12-dihydroxyicosa-6,8,10,14-tetraenoic acid |
Molecular Weight (g/mol) | 336.472 |
PubChem CID | 5280492 |
ChEBI | CHEBI:15647 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'