Learn More
Eppendorf™ Easypet™ 3
Experience of a new dimension of electronic pipetting with this pipette controller. Eppendorf Easypet™ 3 offers intuitive and convenient speed adjustment. Ergonomic design allows fatigue-free pipetting.
Brand: Eppendorf™ 4430000018
485.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Lightweight and well-balanced
- Vibrant backlit LEDs provide optical feedback of the remaining battery life
- Lithium polymer rechargeable battery offers long cordless runtime
- Operation while recharging is possible
- Autoclavable pipette adapter for sterile applications
- Quick release of aspirating cone for easy exchange of membrane filters
Specifications
Lithium-ion | |
Electronic Pipette | |
Membrane |