Learn More
Labconco™ Lid for FreeZone Freeze Dryer
Replacement lid for FreeZone Freeze Dryer
Brand:  Labconco™ 7532900
Product Code. 17281134
  • £112.00 / Each
Estimated Shipment: 30-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Replacement lid for FreeZone Freeze Dryer.
  • 3/4 in. lid, 14 in. diameter lid fits on drying chambers and FreeZone collector chambers.
Specifications
Specifications
Lid
-105°C to +170°C
0.75 in.
14 in.
Up to 18 L
Acrylic
19.05 mm
355.6 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Labconco™ Lid for FreeZone Freeze Dryer > Acrylic