Learn More
Fisherbrand™ Parts/Accessories for OmniPAGE Mini Vertical Electrophoresis System
External casting system (upstand and base), spare tank or spare lid. Fisherbrand™ Parts/Accessories for OmniPAGE Mini Vertical Electrophoresis System are used for applications involving the separation of proteins and nuclueic acids in poly-acrilamide gels.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.01000kg
Description
OmniPAGE Mini Vertical systems are used primarily for protein but also nucleic acid electrophoresis techniques. By combining both functionality and ease of use, the OmniPAGE CVS10 systems enable simple, versatile vertical mini gel electrophoresis with reproducible results.
Each OmniPAGE system can accommodate up to 4 hand-cast gels and 2 commercial precast gels to provide complete flexibility. Unique sliding clamp technology within the PAGE insert facilitates fast, intuitive leak-free casting.
- Electrophoresis Tank: Holds the gel module with cast gels/samples and the buffer solution; connects to an external power source
- Safety Lid: Ensures the correct polarity and orientation of electrodes; fits easily onto tank
- External Casting System: includes upstand and base with silicone mat; securely holds gel module while casting gel
Specifications
Blue | |
OmniPAGE Mini CVS10 Vertical System | |
8 x 8.5 cm | |
Up to 2 |
Protein Gel Electrophoresis | |
Lid (without cables) | |
Without cables | |
10 x 10 cm (plate) |