missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Life Science Sealing Films for PCR and Storage
Detachable, peelable sealing films ideal for PCR, ELISA and sample storage.
Brand: BRAND™ 701376
This item is not returnable.
View return policy
Description
- Due to its adhesive, film can be used down to -80°C
- DMSO-resistant and resistant to common solvents
- Can be removed without leaving any residue so that work can continue unhindered after transport or storage
Specifications
Clear | |
Polyester | |
Not Autoclavable | |
17.2 cm | |
No | |
Round |
Sealing film, PET, peelable | |
Non-sterile | |
96 | |
9 cm | |
-80°C to +120°C | |
PCR Ready |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction