Learn More
Applied Biosystems™ Calibration Kit, Fast 96-well
Used to maintain your real-time PCR instrument with Fast 96-well block.
Brand: Applied Biosystems™ 4432563
Includes: Region of interest (ROI) plate, Background plate, Dye calibration plates, and RNase P instrument verification plate
Description
The kit includes region of interest (ROI), background, and dye calibrations, as well as RNase P verification. Please see your instrument user guide for full instructions on use.
See instrument user guide for full instructions.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Region-of-Insert (ROI) | |
ViiA™ 7 Dx System, QuantStudio™ Dx | |
Real Time PCR (qPCR) | |
Spectral Calibration |
Proprietary Mix | |
Calibration Kit | |
1 x 96-well plate |
Product Content Correction
Product Title
Applied Biosystems™ Calibration Kit, Fast 96-well > 1 x 96-well Plate
For in vitro Diagnostic Use.
Spot an opportunity for improvement?Share a Content Correction