Learn More
Invitrogen™ iBind™ Western Starter Kit
Description
Includes
iBind Western Device, 10 iBind Cards, iBind Solution Kit
The iBind Western Starter Kit includes an iBind Western Device as well as all of the consumables and reagents necessary to get started using this automated Western-processing device that performs every step in the Western protocol from blocking to washes to antibody incubations via sequential lateral flow (SLF). SLF allows the timely release and flow of solutions and antibodies to the membrane without need of an external power source. Each solution is released from the iBind device wells to an iBind Card via SLF; the solutions are then wicked towards the stack region of the card. The glass fiber matrix of the card allows for homogenous and consistent flow of the solutions to the membrane, increasing the antigen-antibody interaction.
- Simple, automated solution
- No external power source
- Superior reproducibility over traditional manual methods
- Better antigen antibody interaction—solutions flow between the iBind Card and membrane for maximum interaction between protein and antibody
- Minimizes hands-on time
- Highly reproducible and compatible with all Western protocols
- Set up time is fast—ready for the detection step within 2.5 hr.
- Carry out detection right away or wait until the next day without affecting performance
- Works exclusively with iBind Cards and the iBind Solution Kit to process one mini-membrane for immunodetection
- Provides automated convenience and superior reproducibility over traditional manual methods
Kit components may also be purchase separately.
Proteins, Expression, Isolation and Analysis, Western Blotting
Specifications
Specifications
| Product Type | Starter Kit |
| Capacity | 1 mini blot (8 x 8 cm) |
| Content And Storage | • 1 iBind™ Western Device • 10 iBind™ Cards (store at room temperature) • 1 iBind™ Solution Kit (store at 2–8°C) |
| Run Time | ∼2.5 hours |
| Detection Method | Chromogenic, Chemiluminescence, Fluorescence |
| Membrane Compatibility | Nitrocellulose, PVDF |
| Product Line | iBind |
| Quantity | 1 kit |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
