Learn More
Invitrogen™ iBind™ Window Cover
Product Code. 15384825
Click to view available options
Quantity:
1 each
Unit Size:
Each
Product Code. 15384825
Brand: Invitrogen™ SLF1001
Only null left
Add to basket
Add to basket
Accessory for the iBind Western Device
Cover fits over the viewing window to provide protection from light during fluorescent detection applications and serve as an indicator for when the device is in use.
Proteins, Expression, Isolation and Analysis, Western Blotting
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Western Blot
For Use With (Equipment) iBind
Product Line iBind
Type Window Cover
Quantity 1 each