missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Standard Calibration Install Kit, 96-well

This kit is used to maintain your ViiA™ 7 System with 96-well Block.

Brand:  Applied Biosystems™ 4432488

985.50 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 12343583

  • £2190.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes:
  • 1 background calibration plate
  • 1 ROI calibration plate
  • 6 spectral calibration plates with FAM™
  • SYBR™ Green I
  • VIC™ NED™ TAMRA™ and ROX™ dye standards
  • 2 normalization calibration plates
  • 1 RNase P instrument verification plate
This item is not returnable. View return policy

Description

Description

It includes calibration plates to perform a spectral calibration with FAM™, VIC™, ROX™, SYBR™ Green, TAMRA™, NED™ dyes, plates to perform region-of-interest (ROI) calibration, normalization calibration, and to run an RNase P verification. Calibration and verification should be run at least every six months. Please see your instrument maintenance guide for full instructions on use. The dye formulations for your ViiA™ 7 System will improve results with multiplexing by more accurately representing fluorescent spectra found in your real-time PCR experiments.

Calibration and verification should be done at least every six months. Please see instrument maintenance guide for full instructions.

Order Info

Store kit at -5° to -30°C

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Includes:
• 1 background calibration plate
• 1 ROI calibration plate
• 6 spectral calibration plates with FAM, SYBR™ Green, VIC, NED, TAMRA, and ROX dye standards
• 2 normalization calibration plates
• 1 RNase P instrument verification plate.

Store kit at -5 to -30°C.
Calibration Install Kit
1 kit
QuantStudio™ 12 K Flex, QuantStudio™ 6 Flex, QuantStudio™ 7, ViiA™ 7 System
Real Time PCR (qPCR)
Spectral Calibration
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.