Learn More
Applied Biosystems™ ViiA™ 7 Array Card Spectral Calibration Kit
Perform spectral calibration, region-of-interest (ROI) calibration and normalization on the ViiA™ 7 or QuantStudio™ 7 Flex or 12K Flex systems with array card block. Applied Biosystems™ ViiA™ 7 Array Card Spectral Calibration Kit includes empty array cards and seven tubes of calibration dyes.
Brand: Applied Biosystems™ 4432314
1642.50 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Includes: Empty array cards and seven tubes of calibration dyes including: FAM™ dye, VIC™ dye, ROX™ dye, ROI dye mix, background buffer, FAM/ROX dye, VIC/ROX dye
Description
It includes empty array cards and seven tubes of calibration dyes including: FAM™ dye, VIC™ dye, ROX™ dye, ROI dye mix, background buffer, FAM™/ROX™ dye, VIC™/ROX™ dye. Please see your instrument maintenance guide for full instructions on use. The dye formulations for your ViiA™ 7, QuantStudio™ 7, or QuantStudio™ 12K Flex system will improve results with multiplexing by more accurately representing fluorescent spectra found in your real-time PCR experiments.
The Applied Biosystems™ ViiA™ 7 Array Card Spectral Calibration Kit includes empty array cards and seven tubes of calibration dyes including: FAM™ dye, VIC™ dye, ROX™ dye, ROI dye mix, background buffer, FAM™⁄ROX™ dye, VIC™⁄ROX™ dye.
The dye formulations for the ViiA™ 7, QuantStudio™ 7, or QuantStudio™ 12K Flex system will improve results with multiplexing by more accurately representing fluorescent spectra found in real-time PCR experiments.
Order Info
Store plates at -15° to -25°C
Specifications
384-well Microfluidic Card | |
FAM, ROX, VIC | |
ViiA™ | |
Real Time PCR (qPCR) | |
Spectral Calibration, Region-of-Interest (ROI) Calibration, Normalization |
Store 7 Tubes of Calibration Dyes at -15°C to -25°C | |
QuantStudio™ 12 K Flex, QuantStudio™ 7, ViiA™ 7 System | |
Array Card Spectral Calibration Kit | |
1 kit |
For Research Use Only. Not for use in diagnostic procedures.