Learn More
JULABO™ Lift-Up Bath Cover for PURA™ Water Baths
Lift-up, transparent bath cover for PURA™ Water Bath up to 99.9°C. Protects against water loss through evaporation. The bath cover is made from plexiglass and polycarbonate.
Brand: JULABO™ 9970584
Additional Details : Weight : 2.43000kg
Description
- Accessory for PURA™ Water Bath
- Prevent water loss through evaporation
- Protect samples from contamination
- Made from plexiglass and polycarbonate
Specifications
Lift-up bath cover | |
PURA™ 30 | |
Plexiglass and polycarbonate |