Learn More
JULABO™ Lift-Up Bath Cover for PURA™ Water Baths
Lift-up, transparent bath cover for PURA™ Water Bath up to 99.9°C. Protects against water loss through evaporation. The bath cover is made from plexiglass and polycarbonate.
Brand:  JULABO™ 9970582
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 16286611
  • £220.00 / Each
Estimated Shipment: 14-08-2024
to see stock.
Add to basket
Description
Description
  • Accessory for PURA™ Water Bath
  • Prevent water loss through evaporation
  • Protect samples from contamination
  • Made from plexiglass and polycarbonate
Specifications
Specifications
Lift-up bath cover
PURA™ 14
Plexiglass and polycarbonate