missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIG4 Mouse anti-Human, Clone: 1H6C11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66705-1-IG-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a DNA ligase that joins single-strand breaks in a double-stranded polydeoxynucleotide in an ATP-dependent reaction. This protein is essential for V(D)J recombination and DNA double-strand break (DSB) repair through nonhomologous end joining (NHEJ). This protein forms a complex with the X-ray repair cross complementing protein 4 (XRCC4), and further interacts with the DNA-dependent protein kinase (DNA-PK). Both XRCC4 and DNA-PK are known to be required for NHEJ. The crystal structure of the complex formed by this protein and XRCC4 has been resolved. Defects in this gene are the cause of LIG4 syndrome. Alternatively spliced transcript variants encoding the same protein have been observed.Specifications
LIG4 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P49917 | |
LIG4 | |
LIG4 Fusion Protein Ag3385 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunofluorescence, Western Blot | |
1H6C11 | |
Unconjugated | |
LIG4 | |
DNA ligase 4, DNA ligase IV, LIG4, ligase IV, DNA, ATP dependent | |
Mouse | |
Protein G | |
RUO | |
3981 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction