missing translation for 'onlineSavingsMsg'
Learn More

Analytik Jena Replacement Bulb with 7° Beam Width for High Intensity UV Lamps

Replacement Bulb for High Intensity UV Lamps with vented housing protects against burns.

Brand:  Analytik Jena 97600-19

Product Code. 12615750

  • £227.00 / Each
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For High Intensity UV Lamps 97600-00, 97600-05, 97600-10, and 97600-15 manufactured after June 2002.
  • These high-intensity lamps produce 365 nm UV light.
  • 1 month warranty.
  • Lamp head rotates 360° in its transformer base.
  • A filter minimizes white light interference allowing you to easily detect weak fluorescence.
  • Detach lamp head from transformer base for portability.
Specifications

Specifications

Light Bulb
100 W
High Intensity UV Lamps
UV
365 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Analytik Jena Replacement Bulb with 7° Beam Width for High Intensity UV Lamps > UV Replacement Bulb; Wavelength: 365 nm; Wattage: 100 W

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.