Learn More
Invitrogen™ EVOS™ Light Cube, AO Red
Brand: Invitrogen™ AMEP4671
This item is not returnable.
View return policy
Description
- Contains unique, proprietary LED light cubes
- Image fluorescent samples for specific wavelengthLight engine outputs high intensity over a short light path that delivers superior fluorophore excitation
- Each cube contains an LED, condensing optics, and hard-coated filters
- User-changeable and automatically recognized by system
- Shorter light path that affords superior detection of fluorescent signals
- Continuous illumination that enables consistent results
- 50,000+ hour bulb lifetime that affords lower operating costs
- Adjustable light intensity that helps reduce photobleaching
Specifications
Light Cube | |
Countess™ II FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System | |
Ex: 442/46 Em: 628/32 | |
1 Ea. |
EVOS | |
Condensing Optics, Hard-Coated Filters, LED | |
Metal Halide (Bulb) |
Product Content Correction
Product Title
Invitrogen™ EVOS™ Light Cube, AO Red > AO Red; Excitation/Emission: 442/46nm, 628/32nm