Learn More
Manutan Light duty mini-roller bearing alloy track, 30 roller bearings, P50, 400mm wide
Product Code. 12875588
Encompass_Preferred
Product Code. 12875588
Brand: Manutan A341701
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Lightweight aluminum gravity conveyor track with PVC roller bearings and mild steel bearings for parcels and packets as small as 70mm in length. Frame: 50mm x 25mm x 3mm aluminum angle, roller bearings: PVC 20mm/30mm diameter. Load capacity per metre: 25kg.
  • 30mm diameter roller bearings with mild steel bearings and spindles
  • Track width: 400mm
  • Roller bearing pitch: 50mm