Learn More
Leica Microsystems Cold Light Source, CLS 100 X
Powerful and economical cold light source. Leica Microsystems™ Cold Light Source, CLS 100 X comes with a universal voltage adapter.
Brand: Leica Microsystems 30250002
This item is not returnable.
View return policy
Description
- Powerful and economical.
- Uniform illuminated area with constant color temperature.
- Continuous brightness control via mechanical diaphragm.
- Exceptionally long bulb service life.
- Fan-cooled light source.
Specifications
Light source | |
Universal adapter |