missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIGHT/TNFSF14 Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHTNFSF14X10

Product Code. 17728043

  • £4370.00 / Pack of 10
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

O43557
15 pg/mL
ELISA
Colorimetric Microplate Reader
TNFSF14
<10%
HRP
RUO
2°C to 8°C
1 hr 20 min
8.23 to 6000 pg/mL
Biotin
Plasma, Serum, Supernatant
8740
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
10 x 96 Tests
Plasma, 50 μL|Serum, 50 μL|Supernatant, 100 μL
LIGHT
4 hr 45 min
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers