missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Lightnin High-Efficiency Axial Flow Impeller

High-efficiency axial flow impellers have laser designed paddles to develop 50% more mixing action for high-flow, low-shear mixing.

Brand:  Cole-Parmer™ 04560-25

Product Code. 15446133

  • £288.00 / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Highly efficient axial flow
  • Paddles are laser disgned - 50% more mixing action
  • High flow, low-shear mixing
Specifications

Specifications

Axial flow impeller
64 mm
8 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.