missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Atto565 Antibody Labeling Kit
Applications: Flow Cytometry
£402.00 - £508.00
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
---|---|
Conjugate | Atto 565 |
Product Type | Rapid Atto565 Antibody Labeling Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18058555
|
Novus Biologicals™
351-0030 |
30 μg |
£402.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18728113
|
Novus Biologicals™
351-0010 |
300 μg |
£508.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Atto565 is one of a new generation of fluorescent labels. It has a strong absorption at 563nm, high fluorescence at 592nm (extinction coefficient 1.2 x105 cm-1M-1) and quantum yield.Specifications
Store at -20°C. Avoid freeze-thaw cycles. | |
Atto 565 | |
Rapid Atto565 Antibody Labeling Kit |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Lightning-Link (R) Rapid Atto565 Antibody Labeling Kit