missing translation for 'onlineSavingsMsg'
Learn More

LIMK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen LIMK1
Concentration 0.17 mg/mL
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16899174
View Documents
Proteintech
19699-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16889174
View Documents
Proteintech
19699-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The antibody has no cross-reaction with LIMK2.

There are approximately 40 known eukaryotic LIM proteins, so named for the LIM domains they contain. LIM domains are highly conserved cysteine-rich structures containing 2 zinc fingers. Although zinc fingers usually function by binding to DNA or RNA, the LIM motif probably mediates protein-protein interactions. LIM kinase-1 and LIM kinase-2 belong to a small subfamily with a unique combination of 2 N-terminal LIM motifs and a C-terminal protein kinase domain. LIMK1 is likely to be a component of an intracellular signaling pathway and may be involved in brain development. LIMK1 hemizygosity is implicated in the impaired visuospatial constructive cognition of Williams syndrome.
Specifications

Specifications

LIMK1
Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Rat, Human, Mouse
P53667, P53668, P53669
16885, 3984, 65172
Peptide
Primary
-20°C
LIMK1
0.17 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
LIM domain kinase 1, LIMK, LIMK 1, LIMK1
LIMK1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications