missing translation for 'onlineSavingsMsg'
Learn More

(+)-Limonene, 96%, unstabilized, Thermo Scientific Chemicals

Catalog Number 10387640
Click to view available options
Quantity:
5 mL
500 mL
2.5 L
Packaging:
Glass Bottle
5989-27-5, 10191-41-0
C10H16
136.24
MFCD00062991
XMGQYMWWDOXHJM-JTQLQIEISA-N
d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene
440917
CHEBI:15382
CC(=C)[C@@H]1CCC(C)=CC1
5989-27-5, 10191-41-0
C10H16
136.24
MFCD00062991
XMGQYMWWDOXHJM-JTQLQIEISA-N
d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene
440917
CHEBI:15382
CC(=C)[C@@H]1CCC(C)=CC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point -74.0°C
Color Colorless to Yellow
Density 0.8405g/mL
Boiling Point 175.4°C to 176.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4700 to 1.4740
Quantity 500 mL
UN Number 2052
Beilstein 05, 133
Merck Index 14, 5493
Specific Gravity 0.8405
Specific Rotation Condition +106.00 (20.00°C c=1,CH3OH)
Specific Rotation + 106.00
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol
IUPAC Name (4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene
Viscosity 0.846 mPa.s (25°C)
Formula Weight 136.24
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material (+)-Limonene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • (+)-Limonene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'