missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIMP-II (SCARB2) Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHSCARB2X10
Additional Details : Weight : 2.50000kg
Description
High density lipoproteins (HDLs) play a critical role in cholesterol metabolism and their plasma concentrations are inversely correlated with risk for atherosclerosis. SR-BI and SR-BII (previously known as SR-BI.2) are the alternatively spliced products of a single gene. SR-BII differs from SR-BI in that the encoded c-terminal cytoplasmic domain is almost completely different. SR-BII binds HDLs and mediates selective uptake of HDL cholesteryl ester, but with an approximately 4-fold lower efficiency than SR-BI. Nuclease protection assays show SR-BII to be abundant in mouse tissues expressing SR-BI, with SR-BII expression found in liver, adrenal glands, and testes. Although the role of SR-BII is not completely clear, research suggests that it may be a functional HDL receptor. In addition, SR-BII mRNA results from the alternative splicing of SR-BI precursor transcripts with the SR-BII isoform mediating selective transfer of lipid between HDL and cells. The relative expression and functional activities of these two isoforms create a potential means of regulating selective lipid transfer between HDL and cells.Specifications
Q14108 | |
10 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
SCARB2 | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
10.24 to 2500 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
950 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 50 μL|Serum, 50 μL|Supernatant, 100 μL | |
LIMP2 | |
4 hr 45 min |