missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Linalool, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
Linalool is used as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions. as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Linalool is used as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions. as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions.
Solubility
Soluble in water (1.589 g/L at 25°C).
Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Linalool is used as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions. as a scent in 60-80% of perfumed hygiene products and cleaning agents including soaps, detergents, shampoos, and lotions.
Solubility
Soluble in water (1.589 g/L at 25°C).
Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 0.865 |
Boiling Point | 199°C to 200°C |
Flash Point | 76°C (168°F) |
Assay Percent Range | 97% |
Refractive Index | 1.462 |
Quantity | 100 mL |
Beilstein | 1721488 |
Merck Index | 14,5495 |
Solubility Information | Soluble in water (1.589g/L at 25°C). |
Formula Weight | 154.25 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'