missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Linalyl acetate, 95%, synthetic, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless |
Density | 0.9010g/mL |
Boiling Point | 220°C |
Flash Point | 85°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (GC) |
Packaging | Glass bottle |
Linear Formula | (CH3)2=CHCH2CH2C(CH3)(OOCCH3)CH=CH2 |
Refractive Index | 1.4490 to 1.452 |
Quantity | 5 mL |
Show More |
Safety and Handling
Product Identifier
- Linalyl acetate, synthetic
- Warning
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'