missing translation for 'onlineSavingsMsg'
Learn More

Linear alkylbenzenesulfonic acid, 97%, Thermo Scientific Chemicals

Product Code. 11347318
Click to view available options
Quantity:
250 g
1 kg
3 x 1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Dodecyl Benzene Sulfonic Acid can be used in all types of household, industrial and institutional cleaners including laundry, dishwash, carwash, hard surface and general purpose detergents. It can also be utilized as an intermediate in preparation of emulsifiers and wetting and foaming agents. Unneutralized compound is an effective surfactant in acidic products such as liquid bowl, aluminum and dairy cleaners. It is an important raw material and intermediate used in Organic Synthesis, Pharma synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68584-22-5
Molecular Formula C18H30O3S
Molecular Weight (g/mol) 326.495
MDL Number MFCD00147445
InChI Key QJRVOJKLQNSNDB-UHFFFAOYSA-N
Synonym p-3-dodecyl benzenesulphonic acid, c10-c16 alkylbenzenesulfonic acid, 3-4-sulfophenyl dodecane, benzenesulfonic acid, 4-1-ethyldecyl, benzenesulfonic acid, c10-16-alkyl derivs., dsstox_cid_7923, 4-dodecan-3-yl benzenesulfonic acid, c10-c16 alkyl benzene sulfonic acid
PubChem CID 29249
IUPAC Name 4-dodecan-3-ylbenzenesulfonic acid
SMILES CCCCCCCCCC(CC)C1=CC=C(C=C1)S(=O)(=O)O

Specifications

Quantity 1 kg
UN Number UN2586
Solubility Information It is soluble in water. 300,000mg/L at 20°C .
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Linear alkylbenzenesulfonic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'