Learn More
Kartell™ Lining for Glass Tapers
Specially designed to prevent machined glass joints from binding
Brand:  Kartell™ 0104200
Additional Details : Weight : 0.02600kg
Product Code. 15835497
  • £30.25 / Pack of 10
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
  • Conforms to DIN 12242
  • Suitable for foodstuff
Compliance
DIN 12242
Specifications
Specifications
34/35
31 mm
10/pk
PTFE