missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Linoleic acid, 99%

Product Code. 11934961 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
This item is not returnable. View return policy

Product Code. 11934961

Brand: Thermo Scientific Chemicals 215041000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60-33-3 | C18H32O2 | 280.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-33-3
Molecular Formula C18H32O2
Molecular Weight (g/mol) 280.45
InChI Key OYHQOLUKZRVURQ-HZJYTTRNSA-N
Synonym linoleic acid, linolic acid, telfairic acid, cis,cis-linoleic acid, 9z,12z-octadeca-9,12-dienoic acid, linoleate, grape seed oil, cis,cis-9,12-octadecadienoic acid, cis-9,cis-12-octadecadienoic acid, 9z,12z-linoleic acid
PubChem CID 5280450
ChEBI CHEBI:17351
IUPAC Name (9Z,12Z)-octadeca-9,12-dienoic acid
SMILES CCCCCC=CCC=CCCCCCCCC(=O)O

Specifications

Melting Point -5.0°C
Color Colorless to Yellow
Density 0.9000g/mL
Boiling Point 229.0°C to 230.0°C (16.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2H
Refractive Index 1.4687 to 1.4707
Quantity 100 mL
Beilstein 02,IV,1754
Merck Index 15,556
Specific Gravity 0.9
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in diethyl ether,miscible with dimethylformamide,soluble in ethanol,petroleum ether
Formula Weight 280.45
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Linoleic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.