missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIPG Polyclonal Antibody, DyLight™ 488, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522918
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody does not react with mouse endothelial lipase.
LIPG (Lipase G, Endohelial type) is an endothelial lipase which is a novel member of the lipoprotein lipase gene family. LIPG is synthesized by endothelial cells and functions at the site of synthesis. LIPG most likely plays a physiological role in modulating lipoprotein metabolism. Overexpressed LIPG has been shown to reduce HDL cholesterol levels, whereas an LIPG deficiency appears to be effective in elevating HDL cholesterol levels in animals. LIPG has substantial phospholipase activity and may be involved in lipoprotein metabolism and vascular biology. Also, LIPG is designated a member of the TG lipase family by its sequence and characteristic lid region which provides substrate specificity for enzymes of the TG lipase family. LIPG is a carboxyl esterase that hydrolyzes high density lipoproteins, emulsifies triglycerides, is essential for the efficient digestion of dietary fats and can bind to heparin.Specifications
LIPG | |
Polyclonal | |
DyLight 488 | |
LIPG | |
3110013K01Rik; EDL; EL; endothelial cell-derived lipase; Endothelial lipase; endothelial-derived lipase; lipase; lipase G; lipase G, endothelial type; lipase, endothelial; LIPG; lipoprotein lipase H; lipose, endothelial; mEDL; PL; PRO719; UNQ387/PRO719 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9388 | |
4° C, store in dark | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
0.65 mg/mL | |
50mM sodium borate with no preservative | |
Q9Y5X9 | |
LIPG | |
An N-terminal synthetic peptide made to the human endothelial lipase protein sequence. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |