Learn More
Lipoamide, MedChemExpress
MedChemExpress Lipoamide is a coenzyme, which transfer acetyl and hydrogen in Pyruvate deacylation oxidation Process, used for pharmaceuticals.
Brand: Medchem Express HY-B1142
This item is not returnable.
View return policy
Description
Lipoamide is a coenzyme, which transfer acetyl and hydrogen in Pyruvate deacylation oxidation Process, used for pharmaceuticals.Specifications
Lipoamide | |
Light Yellow | |
C8H15NOS2 | |
(±)-α-Lipoamide DL-Lipoamide DL-6,8-Thioctamide | |
O=C(N)CCCCC1SSCC1 | |
205.34 | |
Research |
940-69-2 | |
Research | |
100 mg | |
DMSO : ≥ 50 mg/mL (243.50 mM) ∣H2O : < 0.1 mg/mL (insoluble) | |
205.34 | |
96.04% | |
Solid |