missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipofectamine™ 3000 Transfection Reagent
GREENER_CHOICE

Product Code. 15232475
Click to view available options
Quantity:
0.1 mL
0.75 mL
1.5 mL
15 mL
5 x 1.5 mL
Unit Size:
0.1mL
0.75mL
1.5mL
15mL
7.5mL
This item is not returnable. View return policy

Product Code. 15232475

Brand: Invitrogen™ L3000150

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipofectamine™ 3000 Transfection Reagent leverages our most advanced lipid nanoparticle technology to provide superior transfection performance with improved application outcomes and reproducible results.

Lipofectamine™ 3000 Transfection Reagent leverages our most advanced lipid nanoparticle technology to provide superior transfection performance with improved application outcomes and reproducible results.

This reagent delivers superior transfection efficiency and improved cell viability for the widest range of hard-to-transfect and common cells (e.g., HEK293, HeLa).

Successfully transfect the widest variety of biologically relevant cell types with a reagent that offers:
  • Superior performance—our highest efficiency reagent for difficult-to-transfect cells
  • Improved cell viability—gentle on your cells, with low toxicity
  • Versatile—one reagent for DNA, RNA, and co-transfection

Efficiently Transfect Difficult-to-Transfect Cells
Lipofectamine 3000 reagent is designed to efficiently transfect difficult-to-transfect cells, yielding superior transfection performance across the broadest array of cell types. Successfully transfected cell types include:

Fibroblast (3T3, COS-7)
Myoblast (C2C12, L6 CRL-1458)
Hepatocyte (HepG2, HuH7)
Erythroleukemic cell (K562)
Breast carcinoma (MCF7, Hs578T)
Prostate carcinoma (LNCap)
Lung carcinoma (A549, NCI-H460)
Osteosarcoma (U-2 OS, Saos-2)
Colon carcinoma (Caco2, SW480)
Pancreatic carcinoma (PANC-1)
Skin melanoma (SK-MEL-28)

Transfect Common Cells Efficiently and Cost-Effectively
For common cell types, Lipofectamine 3000 reagent is more cost-effective than other reagents due to its high concentration and low usage amount. The 1.5mL size provides up to 1500 transfection reactions (in 24-well plates).

Achieve Superior Transfection Efficiency While Maintaining Low Toxicity
Lipofectamine 3000 reagent maintains a high transfection efficiency within a robust dynamic range of lipid doses for quick and easy optimization. A low toxicity lipid dose (0.75μL for 24-well plates) is suggested for applications requiring minimal disruption of the cells.

Enable High Transfection Efficiency in Novel Genome Editing Applications
Lipofectamine 3000 reagent is suitable for novel genome editing applications. It increases the likelihood of successful cleavage and recombination with TALENs or CRISPRs through highly efficient transfection, ultimately maximizing the efficiency of genetic modifications and simplifying the downstream processes.
TRUSTED_SUSTAINABILITY

Specifications

Classification Animal Origin-Free
Content And Storage Contains one vial of Lipofectamine™ 3000 Reagent and one vial of P3000TM Enhancer Reagent. Store at 2–8°C. Do not freeze.
Cell Type Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Plasmid DNA, Synthetic siRNA, RNAi Plasmids (shRNA, miR)
Transfection Technique Lipid-based Transfection
For Use With (Application) Transfection
Green Features Sustainable packaging
Product Line Lipofectamine
Product Type Transfection Reagent
Quantity 15 mL
Serum Compatible Yes
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.