missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipofectamine™ CRISPRMAX™ Cas9 Transfection Reagent

The first optimized lipid nanoparticle transfection reagent for CRISPR-Cas9 protein delivery.

Brand:  Invitrogen™ CMAX00008

362.07 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15466014

  • £447.00 / 0.75mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Demonstrated cleavage efficiency—tested in over 20 cell types including iPSC, mESC, N2A, CHO, A549, HCT116, HeLa, HEK293, and several others
  • Low cell toxicity—less cells needed to initiate your experiment
  • High throughput friendly—an ideal delivery solution for 96-well format
  • Lipofectamine CRISPRMAX transfection reagent increases the likelihood of successful cleavage and recombination, especially when combined with our TrueCut Cas9 Protein v2 and TrueGuide Synthetic gRNA.
  • Unlike CRISPR plasmid or Cas9 mRNA, using Cas9 protein provides superior cleavage efficiency in primary cells and stem cells.
  • It eliminates the need for transcription or translation of the payload, removes the risk of genomic integration, and is cell cycle independent.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Animal Origin-Free
Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Protein, Ribonucleoprotein, Cas9 Ribonucleoprotein, Cas9, Plasmid DNA
Transfection
Transfection Reagent
Yes
Contains one vial of Lipofectamine CRISPRMAX Reagent and one vial of Cas9 Plus Reagent.

Store at 2–8°C. Do not freeze.
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Lipid-based Transfection
CRISPRMAX™, Lipofectamine™
0.75 mL
Approved for shipment on Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.