missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lipofectamine™ CRISPRMAX™ Cas9 Transfection Reagent

The first optimized lipid nanoparticle transfection reagent for CRISPR-Cas9 protein delivery.

Brand:  Invitrogen™ CMAX00003

143.42 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15435884

  • £202.00 / 25 reactions

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Demonstrated cleavage efficiency—tested in over 20 cell types including iPSC, mESC, N2A, CHO, A549, HCT116, HeLa, HEK293, and several others
  • Low cell toxicity—less cells needed to initiate your experiment
  • High throughput friendly—an ideal delivery solution for 96-well format
  • Lipofectamine CRISPRMAX transfection reagent increases the likelihood of successful cleavage and recombination, especially when combined with our TrueCut Cas9 Protein v2 and TrueGuide Synthetic gRNA.
  • Unlike CRISPR plasmid or Cas9 mRNA, using Cas9 protein provides superior cleavage efficiency in primary cells and stem cells.
  • It eliminates the need for transcription or translation of the payload, removes the risk of genomic integration, and is cell cycle independent.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Animal Origin-Free
Established Cell Lines, Stem Cells, Primary Cells, Hard-to-Transfect Cells
Protein, Ribonucleoprotein, Cas9 Ribonucleoprotein, Cas9, Plasmid DNA
Transfection
Transfection Reagent
Yes
Contains one vial of Lipofectamine CRISPRMAX Reagent and one vial of Cas9 Plus Reagent.

Store at 2–8°C. Do not freeze.
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Lipid-based Transfection
CRISPRMAX™, Lipofectamine™
0.3 mL
Approved for shipment at Room Temperature or on Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.