missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lipofectamine™ MessengerMAX™ Transfection Reagent

Lipofectamine™ MessengerMAX™ Transfection Reagent
£96.75 - £10220.00
Specifications
Content And Storage | Store in refrigerator (2–8°C). |
---|---|
Cell Type | Established Cell Lines, Neuronal Cells, Stem Cells, Primary Cells, Hard-to-Transfect Cells |
Format | 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks |
Sample Type | mRNA |
For Use With (Application) | Transfection |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15321734
![]() |
Invitrogen™
LMRNA001 |
0.1 mL |
£96.75
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15397974
![]() |
Invitrogen™
LMRNA003 |
0.3 mL |
£389.00
0.30mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15364584
![]() |
Invitrogen™
LMRNA008 |
0.75 mL |
£636.00
0.75mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15361844
![]() |
Invitrogen™
LMRNA015 |
1.5 mL |
£1074.00
1.50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15374584
![]() |
Invitrogen™
LMRNA150 |
15 mL |
£10220.00
15mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lipofectamine™ MessengerMAX mRNA Transfection Reagent delivers amazing transfection efficiency in neurons and a broad-spectrum of primary cells, enabling improved application outcomes and more biologically relevant research. That's because our novel lipid nanoparticle technology is optimized to deliver the highest amount of mRNA possible without the nuclear entry step that is required with DNA.Successfully transfect more predictive cell models with a reagent that provides:
Enable high transfection efficiency in novel genome editing applications
Lipofectamine™ MessengerMAX™ reagent helps increase the likelihood of successful cleavage and recombination with GeneArt™ CRISPR Nuclease mRNA through highly efficient transfection, ultimately maximizing the efficiency of genetic modifications and simplifying the downstream processes.
Efficiently transfect difficult-to-transfect cells
Lipofectamine™ MessengerMAX™ reagent is designed to transfect a higher amount of mRNA into neurons and a broad spectrum of difficult-to-transfect primary cells. This eliminates the need for electroporation or viruses, and provides a > 2-fold improvement in transfection efficiency compared to other lipid based reagents.
Leverage advantages of mRNA transfection
Transfection of mRNA does not require nuclear uptake, which results in faster protein expression than DNA transfection since translation of mRNA occurs in the cytoplasm. Also, mRNA has no risk of genomic integration and transfection efficiency is cell cycle independent.
Specifications
Store in refrigerator (2–8°C). | |
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
Transfection | |
Transfection Reagent |
Established Cell Lines, Neuronal Cells, Stem Cells, Primary Cells, Hard-to-Transfect Cells | |
mRNA | |
Sustainable packaging | |
Yes |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Lipofectamine™ MessengerMAX™ Transfection Reagent