missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lipofectamine™ Transfection Reagent
Lipofectamine™ Transfection Reagent
Brand: Invitrogen™ 18324010
257.45 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.01000kg
Description
Lipofectamine™ Transfection Reagent is one of our first-generation, cationic-lipid transfection reagents formulated for the transfection of DNA into eukaryotic cells. Lipofectamine™ Transfection Reagent is a trusted, value reagent with many years of cited use in thousands of publications and with many cell lines.With Lipofectamine™ Transfection Reagent, you'll get:
A proven technology
Our Lipofectamine™-brand reagents have been recognized as the most-cited transfection reagent family, with tens of thousands of citations since launch in 1993. Lipofectamine™ Transfection Reagent is no exception as a trusted reagent for a wide range of cell lines. When used in combination with PLUS™ Reagent, Lipofectamine™ Transfection Reagent has been shown to work well with cells such as BHK-21, NIH 3T3, COS-1, fibroblasts, keratinocytes, HT-29, MRC-5, and SK-BR3.
A 'value' alternative for plasmid DNA
With such a legacy comes a commitment to providing the best reagents for all scientists and labs, regardless of their funding capabilities. Lipofectamine™ Transfection Reagent is a value alternative for transfection of plasmid DNA into eukaryotic cells with a simplified protocol. For optimal results in a wider range of plasmid DNA transfection conditions, we recommend Lipofectamine™ LTX Reagent, due to its optimal balance of potency & low-cytotoxicity. Lipofectamine™ 2000 Transfection Reagent is recommended if you are transfecting a broad range of cell lines or want to transfect RNA.
Specifications
Lipofectamine™ Transfection Reagent, 0.3mL, Liquid, Plasmid DNA Sample Type, Lipid-Based Transfection Technique, No Water Soluble, Used in RNAi, Transfection, Stable Under Normal Conditions, 2 to 8°C Storage Temperature | |
Plasmid DNA | |
RNAi, transfection | |
Lipofectamine™ Transfection Reagent |
Vial | |
Liquid | |
Each unit contains one or multiple vials of reagent | |
0.3 mL |