Learn More
α-Lipoic Acid, MedChemExpress
MedChemExpress α-Lipoic Acid is an antioxidant, which is an essential cofactor of mitochondrial enzyme complexes. α-Lipoic Acid inhibits NF-κB-dependent HIV-1 LTR activation. α-Lipoic Acid induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells.
Brand:  Medchem Express HY-N0492
Encompass_Preferred
Product Code. 16402386
  • £68.97 / 500mg
Explore more special offers
Description
Description
α-Lipoic Acid is an antioxidant, which is an essential cofactor of mitochondrial enzyme complexes. α-Lipoic Acid inhibits NF-κB-dependent HIV-1 LTR activation. α-Lipoic Acid induces endoplasmic reticulum (ER) stress-mediated apoptosis in hepatoma cells.
Specifications
Specifications
α-Lipoic Acid
Yellow
C8H14O2S2
Thioctic acid (±)-α-Lipoic acid DL-α-Lipoic acid
O=C(O)CCCCC1SSCC1
206.33
Research
Cancer-Kinase/protease
1077-28-7
Research
500 mg
DMSO : ≥ 100 mg/mL (484.66 mM) ∣H2O : 0.1 mg/mL (0.48 mM; Need ultrasonic)
206.33
97.0%
Solid