missing translation for 'onlineSavingsMsg'
Learn More

KNF Neuberger™ SIMDOS™ 10 Remote Control Liquid Dosing Pump

Use this pump for a variety of laboratory bench applications. KNF™ SIMDOS™ 10 Remote Control Liquid Dosing Pumps provide safe, precise transfer of even the most aggressive liquids.

Brand:  KNF Neuberger™ 167527/160160

Product Code. 12908074

This item is not returnable. View return policy

Description

Description

Models are available with a choice of pump head materials and in both manual (S) and external control variants (RC).
Chemically resistant diaphragm dosing pump
  • Quick and easy calibration
  • Dosing function (1mL to 1,000mL)
  • Adjustable flow rate (1mL to 100mL/min)
  • Dry running
  • Self-priming
  • Outstanding chemical resistance (FT versions)
For simple dosing of chemicals and other fluids in a variety of applications.
Specifications

Specifications

IP65
PVDF
140 mm
80°C
100/240 V
6 bar
50/60 Hz
SIMDOS™ 10
87 mm
5°C
1 to 100 mL/min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
KNF Neuberger™ SIMDOS™ 10 Remote Control Liquid Dosing Pump > 90-264V 50/60Hz 140mm x 87mm x 130mm flow rate 1 to 100mL/min, volumes 0 to 1000mL PVDF head KNF

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.