Learn More
Agilent BioTek Liquid Handling Control Software
Liquid Handling Control Software provides a powerful yet flexible interface for use with washers, dispensers and combination liquid handling systems.
£1765.00 - £2945.00
Specifications
For Use With (Equipment) | Agilent BioTek Microplate Washers, Dispensers, and Liquid Handling Systems |
---|---|
Type | Liquid Handling Control Software |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
17844667
|
Agilent BioTek
LHC3 |
E-delivery, 5 Installation Licenses |
£1765.00
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17854677
|
Agilent BioTek
LHC3SECURE |
E-delivery, 5 Installation Licenses, 21CFR Part 11 Compliance |
£2945.00
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- LHC software makes it easy to program, even the most complex protocols with a choice of deployment
- Protocols can be run directly from the computer or downloaded to the instrument for convenient onboard recall and operation
- Full instrument control
- For use with 405 TS Washer, 405 LS Washer, ELx405 Select Deep Well Washer, 50 TS Washer, MultiFlo Dispenser, MultiFlo FX Dispenser, EL406 Washer Dispenser, 406 FX Washer Dispenser
- Batch processing with connected BioStack Microplate Stacker or BioSpa 8 Automated Incubator
- Seamless robotic integration to commercially available robotic systems
Specifications
Agilent BioTek Microplate Washers, Dispensers, and Liquid Handling Systems | |
Liquid Handling Control Software |