Learn More
Agilent BioTek Liquid Handling Control Software
GREENER_CHOICE
Offers dual mode control - runs protocols from PC or instrument keypad
Brand:  Agilent BioTek LHC2
Product Code. 15258768
  • £1967.00 / Each
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Intuitive StepWise protocol creation with an unlimited number of methods
  • Create and run protocols from PC or download to instrument's keypad
  • Automatic protocol validation for error-free assurance
  • Protocol parameters printed for safe record keeping and GLP compliance
  • Custom maintenance reminders for continued instrument performance in busy laboratories
  • Supports BioStack integrations for high throughput requirements
  • Security enabled protection for downloaded protocols
  • Multiple instruments controlled in parallel