missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Liquid Master

Available in a range of lengths and diameters to give a range of sample volumes. Sampling Systems™ Liquid Master is ideal for taking samples from low and medium viscosity liquids.

Brand:  Sampling Systems™ 5035P-600

Product Code. 15839331

  • £306.00 / Each
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Product Contact Material Available in: 316L Stainless Steel, Food Grade PTFE/FEP, Polypropylene
  • Easily stripped down for cleaning
  • Single Point and Cross Sectional Sampling
Specifications

Specifications

Liquid Sampler
150 mL
25 mm
Polypropylene
Sampling Liquids
600 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers