missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Liquid Nitrogen Secondary Trap
For use with processing samples with ultra low eutectic points when lyophilizing. Labconco™ Liquid Nitrogen Secondary Trap connects in series with a freeze dry system and vacuum pump to prevent contaminants from migrating into the vacuum pump
Brand: Labconco™ 7394800
4510.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For processing samples with ultra low eutectic points
- Secondary Trap connects in series with a freeze dry system and vacuum pump to prevent contaminants from migrating into the vacuum pump
- After pulling a vacuum, liquid nitrogen may be introduced via the liquid nitrogen port
- Liquid nitrogen cools to -196°C (-321°F) to trap contaminants with low eutectic points
- The well may be defrosted and the liquid drained from the drain hose
- Glacier white powder-coated steel construction
- Two valves may be opened (trap) or closed (by-pass)
- Opened valves allow gaseous contaminants to be trapped in the liquid nitrogen well; Closed valves allow gaseous contaminants to by-pass the well during defrosting and draining
- 3/4 in. (19.05 mm) OD inlet and outlet connections
- Includes an insulated filler tube for connection to a user-supplied liquid nitrogen tank; 20 in. (508 mm) wire-reinforced PVC tubing, 3/4 in. (19.05 mm) ID; and two clamps
- Liquid nitrogen is required (not included)
Specifications
-196°C | |
FreeZone Freeze Dry Systems |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Labconco™ Liquid Nitrogen Secondary Trap > Temperature Range: -196°C
Spot an opportunity for improvement?Share a Content Correction