missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Liquid Proteinase K

Product Code. 15810707
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 15810707

Brand: Macherey-Nagel™ 740396

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Liquid Proteinase K (14-22 mg/mL, > 50 U/mL)

  • Liquid Proteinase K is a recombinant enzyme from Tritirachium album and is expressed in Pichia pastoris
  • Has the identical molecular structure and enzymatic characteristics as the native protein.
  • Very robust endopeptidase that inactivates proteins without any pronounced cleavage specificity
  • Very active in the presence of agents that denature proteins (e.g., SDS and urea)
  • Destructs proteins in cell lysates and promotes the isolation of nucleic acids
  • Rapidly inactivates nucleases, such as DNases and RNase

Specifications

Concentration 14 to 22 mg/mL
Color Colorless
Components Proteinase K liquid, Glycerole
For Use With (Application) RNA/DNA isolation
For Use With (Equipment) NucleoSpin and NucleoMag kits
CAS 39450-01-6l
Content And Storage Room temperature (18-25 °C) for at least one year
Source Tritirachium album expressed in Pichia pastoris
Quantity 5 mL
Form Ready-to-Use Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.