missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Liquid Scintillation Vials, HDPE, 20mL
Made from high density polyethylene with lightweight walls for increased counting efficiency. These LS vials with separate screw caps (size 22-400) are packaged in polyethylene bags of 100 and placed in a separate tray from the caps.
Brand: DWK Life Sciences 986716
This item is not returnable.
View return policy
Description
- Made from high density polyethylene with lightweight walls for increased counting efficiency
- Manufactured to precise tolerances to avoid jamming
- Packaged in 5 utility trays with each partition tray holding 100 vials
- Utility trays serve as a way to store your samples
Specifications
Polyethylene Cone (cap) | |
HDPE | |
500 | |
Urea | |
22-400 |
27 mm | |
20 mL | |
Shrink-wrapped. Caps packaged separately. | |
61 mm |